missing translation for 'onlineSavingsMsg'
Learn More

TRIM35 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA540889

Code : NEW

Additional Details : Weight : 0.00010kg

Product Code. 15534472

  • £404.00 / 100µL
Estimated Shipment: 08-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Peptide sequence: ATKQLCKEFT DLLAQDRTPI GNSRPSPILE PGIQSCLTHF SLITHGFGAP Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 79%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 100%; Rabbit: 93%; Rat: 92%

TRIM35 Polyclonal antibody specifically detects TRIM35 in Human samples. It is validated for Immunohistochemistry, Western Blot.
Specifications

Specifications

TRIM35
Polyclonal
Unconjugated
TRIM35
0710005M05Rik; A430106H13Rik; AW046487; Hemopoietic lineage switch protein 5; HLS5; KIAA1098; LOW QUALITY PROTEIN: tripartite motif-containing protein 35; macrophage-derived apoptosis-inducing RBCC protein; Mair; MGC17233; mKIAA1098; NC8; Protein MAIR; Protein Nc8; Trim35; tripartite motif containing 35; tripartite motif protein 35; tripartite motif-containing 35; tripartite motif-containing protein 35
Rabbit
Affinity Chromatography
RUO
23087
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry, Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9UPQ4
TRIM35
Synthetic peptide directed towards the middle region of human TRIM35
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers