missing translation for 'onlineSavingsMsg'
Learn More

TRAIL/TNFSF10 Rat anti-Mouse, Clone: 170533, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1121

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458365

  • £353.77 / 500µg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

TRAIL/TNFSF10 Monoclonal antibody MAB1121 Specifically detects TRAIL/TNFSF10 in Mouse samples. It is validated for ELISA Capture, Western Blotting
Specifications

Specifications

TRAIL/TNFSF10
170533
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
TNFSF10
E. coli-derived recombinant mouse TRAIL/TNFSF10, Pro118-Asn291, Accession # P50592
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
8743
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, ELISA
LYOPH
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
P50592
Apo-2 ligand, APO2L, Apo-2Ltumor necrosis factor (ligand) family, member 10, APO2Ltumor necrosis factor apoptosis-inducing ligand splice variant delta, CD253, CD253 antigen, Protein TRAIL, TL2, TNF-related apoptosis-inducing ligand, TNFSF10, TRAILTNF-related apoptosis inducing ligand TRAIL, tumor necrosis factor (ligand) superfamily, member 10, tumor necrosis factor ligand superfamily member 10
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects mouse TRAIL/TNFSF10 in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) APRIL, rhBAFF, recombinant mouse (rm) BAFF, rhEDA-A2, rmEDA, rhFas Ligand, rmFas Ligand, rhGITR Ligand, rhLIGHT, rhOX40 Ligand, cotton rat TNF-alpha , rhTNF-alpha , rmTNF-alpha , recombinant porcine TNF-alpha , recombinant rat TNF-alpha , recombinant rhesus macaque TNF-alpha , rhTRANCE, rhTWEAK, rhVEGI, or rhTRAIL.
Mouse
Videos

For Research Use Only