missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF3IP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522801
Code : LD
Additional Details : Weight : 0.01000kg
Description
Suggested positive control: antigen standard for TRAF3IP2 (transient overexpression lysate).
TRAF3IP2 Polyclonal antibody specifically detects TRAF3IP2 in Human, Mouse samples. It is validated for Western Blot.Specifications
TRAF3IP2 | |
Polyclonal | |
Unconjugated | |
TRAF3IP2 | |
ACT1; adapter protein CIKS; AI429613; C6orf2; C6orf4; C6orf5; C6orf6; CANDF8; CIKS; connection to IKK and SAPK/JNK; NFkB-activating protein ACT1; nuclear factor NF-kappa-B activator 1; PSORS13; RGD1304768; RP3-487J7.3; TRAF3 interacting protein 2; TRAF3-interacting protein 2; TRAF3IP2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
103213, 10758 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O43734 | |
TRAF3IP2 | |
Synthetic peptide corresponding to the N-terminus of human ACT. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |