missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517500
Code : LE
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody.
TRAF2 Polyclonal antibody specifically detects TRAF2 in Human, Mouse, Non-human Primate samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
TRAF2 | |
Polyclonal | |
Unconjugated | |
TRAF2 | |
AI325259; E3 ubiquitin-protein ligase TRAF2; MGC:45012; OTTHUMP00000064745; RING-type E3 ubiquitin transferase TRAF2; TNF receptor associated factor 2; Tnf receptor-associated factor 2; TRAF2; TRAP; TRAP3; tumor necrosis factor type 2 receptor associated protein 3; tumor necrosis factor type 2 receptor-associated protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22030, 7186 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
25 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P39429, Q12933 | |
TRAF2 | |
Synthetic peptide corresponding to residues surrounding cysteine 192 of human TRAF2 | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate | |
Antibody | |
IgG |