missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRADD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534807
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: MCF-7, MDA-MB-231. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TRADD Polyclonal antibody specifically detects TRADD in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
TRADD | |
Polyclonal | |
Unconjugated | |
TRADD | |
9130005N23Rik; AA930854; Hs.89862; Hs89862; MGC11078; TNF receptor 1 associated signal transducer; TNFR1-associated DEATH domain protein; TNFRSF1A associated via death domain; TNFRSF1A-associated via death domain; TRADD; tumor necrosis factor receptor type 1 associated death domain protein; tumor necrosis factor receptor type 1-associated DEATH domain protein; tumor necrosis factor receptor-1-associated protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8717 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.85 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q15628 | |
TRADD | |
Recombinant fragment corresponding to a region within amino acids 85 and 312 of TRADD (Uniprot ID#Q15628) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |