Learn More
TRADD Mouse anti-Human, Clone: 313203, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2658
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13449605
  • £277.83 / 100µg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
TRADD Monoclonal antibody MAB2658 Specifically detects TRADD in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Western Blotting
Specifications
Specifications
TRADD
313203
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
TRADD
E. coli-derived recombinant human TRADD Met1-Ala312 Accession # Q15628
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
8717
Reconstitute at 0.5 mg/mL in sterile PBS.
Flow Cytometry, Immunocytochemistry, CyTOF
LYOPH
Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 3-25 ug/mL, CyTOF-ready
Q15628
MGC11078, TNFRSF1A-associated via death domainTNFR1-associated DEATH domain protein, tumor necrosis factor receptor type 1 associated death domain protein, tumor necrosis factor receptor type 1-associated DEATH domain protein, tumor necrosis factor receptor-1-associated protein
Mouse
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human TRADD in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) BAFF R, rhTNF R1, rhHVEM, rhLT beta R, rhCD30, rhGITR, rh4-1BB, rhDR3, rhDR6, rhRANK, rhRELT, rhTAJ, rhEDAR, rhFas, recombinant mouse (rm) DC-TRAIL R1, rmDC-TRAIL R2, rhTRAIL R3, rhTRAIL R4, rhNGF R, rhCD27, rhTNF RII, rhTWEAK R, rhTRAIL R2, or rhCD40 is observed.