Learn More
TRACP/PAP/ACP5 Mouse anti-Human, Clone: 417603, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3948
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13440336
  • £302.82 / 100µg
Call for latest availability
+44 (0)1509 555 500
Description
Description
TRACP/PAP/ACP5 Monoclonal antibody MAB3948 Specifically detects TRACP/PAP/ACP5 in Human samples. It is validated for Western Blotting
Specifications
Specifications
TRACP/PAP/ACP5
417603
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human TRACP in direct ELISAs and Western blots.
Human
Western Blot
LYOPH
Western Blot 1 ug/mL
P13686
ACP5
Mouse myeloma cell line NS0-derived recombinant human TRACP/PAP/ACP5 Ala22-Pro320 Accession # P13686
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
54
Reconstitute at 0.5 mg/mL in sterile PBS.