missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRA-1-60 Monoclonal Antibody (TRA-1-60), HRP, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA1023HRP
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA1-023-HRP has been successfully used in Western blot applications on human samples.
TRA-1-60 Monoclonal antibody specifically detects TRA-1-60 in Human samples. It is validated for Western Blot.Specifications
TRA-1-60 | |
Monoclonal | |
1 mg/mL | |
proprietary buffer with proprietary stabilizer and 0.07% Kathon | |
Mouse | |
Affinity chromatography - MBP | |
RUO | |
Human | |
Antibody | |
IgM |
Western Blot | |
TRA-1-60 | |
HRP | |
TRA160; tra60 | |
Human embryonal carcinoma cell line 2102Ep. | |
100 μL | |
Primary | |
4° C, do not freeze | |
Liquid |