missing translation for 'onlineSavingsMsg'
Learn More

TPT1, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA535332

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15358054

  • £304.30 / 400µL


Estimated Shipment
04-07-2018
Add to basket

Description

Description

This antibody is predicted to react with bovine, porcine and rabbit based on sequence homology.
Specifications

Specifications

TPT1
Polyclonal
PBS with 0.09% sodium azide
P13693
TPT1, Translationally-controlled tumor protein, Fortilin, Histamine-releasing factor, p23
Rabbit
IgG
400μL
Antibody
Polyclonal
Human
Flow CytometryþImmunohistochemistry (Paraffin)þWestern Blot
Unconjugated
TPT1
Purified
TPT1
KLH conjugated synthetic peptide between 85-113 amino acids from the central region of human TPT1
Size-exclusion - Dialysis, Ammonium sulfate precipitation
Store at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
7178
Videos