missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLN2 Monoclonal Antibody (254008), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA523905
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
CLN2 Monoclonal antibody specifically detects CLN2 in Human samples. It is validated for Immunoprecipitation, Western Blot.Specifications
CLN2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O14773 | |
TPP1 | |
Mouse myeloma cell line NS0-derived recombinant human Tripeptidyl-Peptidase I/TPP1 Ser20-Pro563 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunoprecipitation, Western Blot | |
254008 | |
Unconjugated | |
TPP1 | |
cell growth-inhibiting gene 1 protein; ceroid-lipofuscinosis, neuronal 2; CLN 2; CLN2; GIG 1; GIG1; growth-inhibiting protein 1; LPIC; lysosomal pepstatin insensitive protease; lysosomal pepstatin-insensitive protease; MGC21297; SCAR7; TPP 1; TPP I; Tpp1; TPP-1; TPPI; TPP-I; Tripeptidyl aminopeptidase; tripeptidyl peptidase 1; tripeptidyl peptidase I; tripeptidyl peptidase-I; tripeptidyl-peptidase 1; Tripeptidyl-peptidase I; UNQ267/PRO304 | |
Mouse | |
Protein A/G | |
RUO | |
1200 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |