Learn More
TORC1 Rat anti-Mouse, Clone: 505531, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB5978
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13451166
  • £286.65 / 100µg
Estimated Shipment date
from Supplier 03-04-2020
Description
Description
TORC1 Monoclonal antibody MAB5978 Specifically detects TORC1 in Mouse samples. It is validated for Immunohistochemistry
Specifications
Specifications
TORC1
505531
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CRTC1
E. coli-derived recombinant mouse TORC1 Gln384-Gln471 Accession # Q68ED7
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
23373
Reconstitute at 0.5 mg/mL in sterile PBS.
Immunohistochemistry
LYOPH
Immunohistochemistry 8-25 ug/mL
Q68ED7
CREB regulated transcription coactivator 1, CRTC1, FLJ14027WAMTP1, KIAA0616Transducer of regulated cAMP response element-binding protein 1, MECT1, MECT1CREB-regulated transcription coactivator 1, mucoepidermoid carcinoma translocated 1, Mucoepidermoid carcinoma translocated protein 1, TORC1TORC-1, Transducer of CREB protein 1, transducer of regulated cAMP response element-binding protein (CREB) 1, WAMTP1
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse TORC1 in direct ELISAs.