missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-TNFRSF13B, Clone: 4C4, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00023495-M09.100ug
Additional Details : Weight : 0.02000kg
Specifications
TNFRSF13B | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_012452 | |
TNFRSF13B | |
TNFRSF13B (NP_036584.1, 2 a.a. ∼ 165 a.a) partial recombinant protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
4C4 | |
Mouse monoclonal antibody raised against a partial recombinant TNFRSF13B. | |
TNFRSF13B | |
CD267/CVID/FLJ39942/MGC133214/MGC39952/TACI/TNFRSF14B | |
Mouse | |
Affinity chromatography | |
RUO | |
23495 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
For Research Use Only