Learn More
TNF RII/TNFRSF1B Hamster anti-Mouse, Clone: TR7532, R&D Systems™
Hamster Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB4261
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13440486
  • £337.93 / 500µg
Estimated Shipment
09-07-2019
Description
Description
TNF RII/TNFRSF1B Monoclonal antibody MAB4261 Specifically detects TNF RII/TNFRSF1B in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
TNF RII/TNFRSF1B
TR7532
Western Blot 1 ug/mL
P20333
CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNFRII, TNF-RII, TNF-R-II, TNFR-II, TNFRSF1B, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1B, tumor necrosis factor receptor superfamily, member 1B, Tumor necrosis factor receptor type II
Hamster
IgG
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse TNF RII in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) 4-1BB, rm4-1BB, rhAXL, rhCD27, rmCD27, rhCD30, rmCD30, rhCD40, rmCD40, rhDR3, rhDR6, rhEDAR, rmEDAR, rhFas, rmFas, rhGITR, rmGITR, rhHVEM, rhLymphotoxin-beta R, rmLymphotoxin-beta R, rhNGF R, rhOPG, mOPG, rhRANK, rmRANK, rhTAJ, rhTNF RI, rmTNF RI, rhTNF RII.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
TNFRSF1B
E. coli-derived recombinant mouse TNF RII Extracellular domain Accession # P20333
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
7133
Reconstitute at 0.5 mg/mL in sterile PBS.