Learn More
TNF RII/TNFRSF1B Hamster anti-Mouse, Clone: TR75-89.29, R&D Systems™
Hamster Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB4262
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13460486
  • £337.93 / 500µg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
TNF RII/TNFRSF1B Monoclonal antibody MAB4262 Specifically detects TNF RII/TNFRSF1B in Mouse samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
TNF RII/TNFRSF1B
TR75-89.29
Western Blot 1 ug/mL, Immunoprecipitation
Purified
TNFRSF1B
E. coli-derived recombinant mouse TNF RII/TNFRSF1B
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
7133
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNFRII, TNF-RII, TNF-R-II, TNFR-II, TNFRSF1B, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1B, tumor necrosis factor receptor superfamily, member 1B, Tumor necrosis factor receptor type II
Hamster
IgG
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse TNF RII/TNFRSF1B in direct ELISAs and Western blots.