Learn More
TNF alpha Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen P350
Code : PN
Additional Details : Weight : 0.00010kg
Description
P350 targets TNF Alpha in ELISA, IA, and WB applications and shows reactivity with mouse samples. The P350 immunogen is recombinant mouse TNF alpha. P350 detects TNF Alpha which has a predicted molecular weight of approximately 26 kDa. This product is a Low Endotoxin formulation. This product has been tested for endotoxins by limulus amoebocyte lysate (LAL) assay and contains an endotoxin concentration of less than or equal to 10 endotoxin units per milligram (EU/mg).
TNF alpha Polyclonal antibody specifically detects TNF alpha in Mouse samples. It is validated for ELISA, Inhibition Assays, Western Blot.Specifications
TNF alpha | |
Polyclonal | |
Unconjugated | |
Tnf | |
APC1 protein; Cachectin; C-domain 1; C-domain 2; cTNF; DADB-70P7.1; DIF; ICD1; ICD2; Intracellular domain 1; Intracellular domain 2; N-terminal fragment; NTF; RATTNF; Tnf; TNF alpha; TNF superfamily; TNF a; TNF, macrophage-derived; TNF, monocyte-derived; TNFA; TNF-a; TNFalpha; TNF-alpha; Tnfsf1a; TNFSF2; TNFa; TNLG1F; Tumor necrosis factor; tumor necrosis factor (TNF superfamily, member 2); tumor necrosis factor alpha; tumor necrosis factor alpha (cachetin); tumor necrosis factor alpha precursor; tumor necrosis factor ligand 1F; tumor necrosis factor ligand superfamily member 2; Tumor necrosis factor, membrane form; Tumor necrosis factor, soluble form; tumor necrosis factor-alpha; tumor necrosis factor-alpha precursor; tumor-necrosis factor; tumour necrosis factor | |
Rabbit | |
Protein A | |
RUO | |
21926 | |
-20°C | |
Liquid |
ELISA, Inhibition Assays, Western Blot | |
1 mg/mL | |
PBS with no preservative | |
P06804 | |
Tnf | |
Recombinant mouse TNF alpha | |
1 mg | |
Primary | |
Mouse | |
Antibody | |
IgG |