missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF alpha Monoclonal Antibody (MP6-XT3), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen AMC3812
Code : 50
Additional Details : Weight : 0.50000kg
Description
TNF alpha Monoclonal antibody specifically detects TNF alpha in Mouse samples. It is validated for Functional Assay, Neutralization.Specifications
TNF alpha | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.2 | |
P06804 | |
Tnf | |
Recombinant TNF-alpha | |
500 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 κ |
Functional Assay, Neutralization | |
MP6-XT3 | |
Unconjugated | |
Tnf | |
APC1 protein; Cachectin; C-domain 1; C-domain 2; cTNF; DADB-70P7.1; DIF; ICD1; ICD2; Intracellular domain 1; Intracellular domain 2; N-terminal fragment; NTF; RATTNF; Tnf; TNF alpha; TNF superfamily; TNF a; TNF, macrophage-derived; TNF, monocyte-derived; TNFA; TNF-a; TNFalpha; TNF-alpha; Tnfsf1a; TNFSF2; TNFa; TNLG1F; Tumor necrosis factor; tumor necrosis factor (TNF superfamily, member 2); tumor necrosis factor alpha; tumor necrosis factor alpha (cachetin); tumor necrosis factor alpha precursor; tumor necrosis factor ligand 1F; tumor necrosis factor ligand superfamily member 2; Tumor necrosis factor, membrane form; Tumor necrosis factor, soluble form; tumor necrosis factor-alpha; tumor necrosis factor-alpha precursor; tumor-necrosis factor; tumour necrosis factor | |
Rat | |
purified | |
RUO | |
21926 | |
-20°C | |
Liquid |