Learn More
TNF-alpha Rat anti-Rhesus Macaque, Clone: 182312, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB10702
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13458305
  • £337.93 / 500µg
Estimated Shipment
06-03-2019
Add to basket
Description
Description
TNF-alpha Monoclonal antibody MAB10702 Specifically detects TNF-alpha in Rhesus Ma samples. It is validated for ELISA Capture
Specifications
Specifications
TNF-alpha
182312
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
TNF
E. coli-derived recombinant rhesus macaque TNF-alpha Val77-Leu233 Accession # P48094
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
7124
Reconstitute at 0.5 mg/mL in sterile PBS.
ELISA
LYOPH
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
P48094
APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, tnfa, TNF-a, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects rhesus macaque TNF-alpha in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse (rm) EDA, rmFas Ligand, rmOX40, rcrTNF-alpha , rhTNF-alpha , rmTNF-alpha , rpTNF-alpha , rrTNF-alpha , rmTRAIL, or rmTWEAK.
Rhesus Macaque