missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMP21 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA146005
Code : LD
Additional Details : Weight : 0.01000kg
Description
Suggested positive control: human liver protein.
TMP21 Polyclonal antibody specifically detects TMP21 in Bovine, Human, Mouse, Non-human Primate, Rabbit, Rat, X. laevis samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
TMP21 | |
Polyclonal | |
Unconjugated | |
TMED10 | |
1110014C03Rik; 21 kDa transmembrane trafficking protein; 21 kDa transmembrane-trafficking protein; hypothetical protein; integral membrane protein p23; integral membrane protein Tmp21-I (p23); p23; p24 family protein delta-1; P24(DELTA); p24d1; p24delta; p24delta1; S31I125; S31III125; testicular tissue protein Li 206; tmed10; tmed10.L; TMP21; Tmp-21-I; transmembrane emp24 domain-containing protein 10; transmembrane emp24-like trafficking protein 10; transmembrane emp24-like trafficking protein 10 (yeast); transmembrane p24 trafficking protein 10; transmembrane p24 trafficking protein 10 L homeolog; transmembrane protein; transmembrane protein tmp21; transmembrane trafficking protein; transmembrane trafficking protein 21; XELAEV_18000191mg; Xp24d2; Xp24delta2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100009296, 10972, 379442, 529761, 68581, 84599 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.95 mg/mL | |
PBS with 30% glycerol and 0.1% sodium azide | |
P49755, Q28735, Q5E971, Q63584, Q9D1D4 | |
TMED10, tmed10.L | |
Synthetic internal peptide corresponding to residues 50-150 of the human TMP21 protein. | |
100 μL | |
Primary | |
Bovine, Human, Mouse, Non-human Primate, Rabbit, Rat, X. laevis | |
Antibody | |
IgG |