Learn More
TLR9 Mouse anti-Human, Clone: 229106, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3658
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13440196
  • £288.60 / 100µg
Estimated Shipment
19-08-2020
Description
Description
TLR9 Monoclonal antibody MAB3658 Specifically detects TLR9 in Human samples. It is validated for Flow Cytometry (Intracellular Staining)
Specifications
Specifications
TLR9
229106
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
TLR9
NS0 mouse myeloma cell line transfected with human TLR9 Accession # NP_059138
IgM-specific Affinity-purified from hybridoma culture supernatant
RUO
Primary
54106
Reconstitute at 0.5 mg/mL in sterile PBS.
Flow Cytometry, CyTOF
LYOPH
Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
NP_059138
CD289, CD289 antigen, toll-like receptor 9
Mouse
IgM
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human TLR9. Stains human TLR9-transfected cells but not irrelevant transfectants.