missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR6 Monoclonal Antibody (86B1153.2), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516177
Code : LD
Additional Details : Weight : 0.10000kg
Description
TLR6 Monoclonal antibody specifically detects TLR6 in Human samples. It is validated for Flow Cytometry.Specifications
TLR6 | |
Monoclonal | |
0.37 mg/mL | |
PBS with 0.05% sodium azide | |
Q9Y2C9 | |
TLR6 | |
Synthetic peptide corresponding to amino acids 408-424 of human TLR6. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
86B1153.2 | |
PE | |
TLR6 | |
CD antigen 286; CD286; TLR6; toll like receptor 6; toll-like receptor 6 | |
Mouse | |
Protein G | |
RUO | |
10333 | |
4° C, store in dark | |
Liquid |