missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA524246
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with mouse based on sequence homology.
TLL1 Polyclonal antibody specifically detects TLL1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
TLL1 | |
Polyclonal | |
Unconjugated | |
TLL1 | |
ASD6; TLL; TLL1; tolloid like 1; tolloid-like 1; tolloid-like protein 1 | |
Rabbit | |
Protein A | |
RUO | |
7092 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
O43897 | |
TLL1 | |
KLH conjugated synthetic peptide between 874-901 amino acids from the C-terminal region of human TLL1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |