missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RARRES1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530227
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, Raji. Predicted reactivity: Rat (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RARRES1 Polyclonal antibody specifically detects RARRES1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
RARRES1 | |
Polyclonal | |
Unconjugated | |
RARRES1 | |
5430417P09Rik; AI662122; latexin-like; LXNL; PEIG1; PERG-1; phorbol ester-induced gene 1 protein; Rarres1; RAR-responsive protein TIG1; retinoic acid receptor responder (tazarotene induced) 1; retinoic acid receptor responder 1; retinoic acid receptor responder protein 1; Tazarotene-induced gene 1 protein; tazarotene-induced protein 1; TIG1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5918 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P49788 | |
RARRES1 | |
Recombinant fragment corresponding to a region within amino acids 93 and 294 of Human TIG1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |