Learn More
Thymopoietin Mouse anti-Human, Clone: 6E10, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB819
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13461896
  • £337.93 / 500µg
Estimated Shipment
07-03-2019
Add to basket
Description
Description
Thymopoietin Monoclonal antibody MAB819 Specifically detects Thymopoietin in Human samples. It is validated for Western Blotting
Specifications
Specifications
Thymopoietin/LAP2
60000000000
Western Blot 1 ug/mL
P42166
CMD1T, lamina-associated polypeptide 2, LAP2PRO0868, LEM domain containing 4, LEMD4, MGC61508, thymopoietin, Thymopoietin isoform alpha, Thymopoietin, isoforms beta/gamma, Thymopoietin-related peptide isoform alpha, Thymopoietin-related peptide isoforms beta/gamma, TMPO, TP, TP alpha, TP beta/gamma, TPRP isoform alpha, TPRP isoforms beta/gamma
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Thymopoietin in direct ELISAs and Western blots.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
TMPO
E. coli-derived recombinant human Thymopoietin aa 2-187 Accession # P42166
Protein A or G purified from ascites
RUO
Primary
7112
Reconstitute at 0.5 mg/mL in sterile PBS.