missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD90 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA511917
Code : LE
Additional Details : Weight : 0.01000kg
Description
CD90 Polyclonal antibody specifically detects CD90 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
CD90 | |
Polyclonal | |
Unconjugated | |
Thy1 | |
CD7; CD90; CDw90; FLJ33325; membrane glycoprotein; T25; Thy 1.2; THY1; Thy-1; Thy-1 antigen; Thy-1 cell surface antigen; thy-1 glycoprotein; thy-1 membrane glycoprotein; Thy-1 protein; Thy-1 T-cell antigen; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta; Thymus cell surface antigen | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
7070 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P04216 | |
Thy1 | |
KLH conjugated synthetic peptide between 36-65 amino acids from the N-terminal region of human THY1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |