missing translation for 'onlineSavingsMsg'
Learn More

THUMP domain containing 1, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human THUMPD1 protein.

Brand:  Abnova H00055623-B01.50uL

Additional Details : Weight : 0.00010kg

Product Code. 16148246

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Sequence: MAAPAQQTTQPGGGKRKGKAQYVLAKRARRCDAGGPRQLEPGLQGILITCNMNERKCVEEAYSLLNEYGDDMYGPEKFTDKDQQPSGSEGEDDDAEAALKKEVGDIKASTEMRLRRFQSVESGANNVVFIRTLGIEPEKLVHHILQDMYKTKKKKTRVILRMLPISGTCKAFLEDMKKYAETFLEPWFKAPNKGTFQIVYKSRNNSHVNREEVIRELAGIVCTLNSENKVDLTNPQYTVVVEIIKAVCCLSVVKDYMLFRKYNLQEVVKSPKDPSQLNSKQGNGKEAKLESADKSDQNNTAEGKNNQQVPENTEELGQTKPTSNPQVVNEGGAKPELASQATEGSKSNENDFS
Specifications

Specifications

THUMP domain containing 1
Polyclonal
Mouse polyclonal antibody raised against a full-length human THUMPD1 protein.
THUMPD1
DKFZp686C1054
Mouse
50 μL
Yes
55623
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Immunofluorescence, Western Blot
Unconjugated
No additive
NM_017736.3
THUMPD1
THUMPD1 (NP_060206.2, 1 a.a. ∼ 353 a.a) full-length human protein.
RUO
Primary
Human
Serum
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers