Learn More
TARS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530690
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (98%), Rat (98%), Zebrafish (93%), Xenopus laevis (93%), Chicken (91%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TARS Polyclonal antibody specifically detects TARS in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
TARS | |
Polyclonal | |
Unconjugated | |
TARS1 | |
D15Wsu59e; hypothetical protein LOC510075; MGC9344; TARS; TARS1; threonine tRNA ligase 1, cytoplasmic; threonine--tRNA ligase; threonine--tRNA ligase 1, cytoplasmic; threonine--tRNA ligase 1, cytoplasmic; threonine--tRNA ligase, cytoplasmic; threonine--tRNA ligase, cytoplasmic; Threonyl-tRNA synthetase; threonyl-tRNA synthetase 1; threonyl-tRNA synthetase, cytoplasmic; ThrRS; wu:fb07c05; wu:fb39c12; zgc:92586; zgc:92586 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
110960, 6897 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P26639, Q9D0R2 | |
TARS1 | |
Recombinant fragment corresponding to a region within amino acids 346 and 657 of Human ThrRS | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |