missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THSD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543330
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: AQGQWVEFGC APLGPEAYVT VVLKLLGRDS VITSTGPIDL AQKFGYKLVM Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%
THSD1 Polyclonal antibody specifically detects THSD1 in Human samples. It is validated for Western Blot.Specifications
THSD1 | |
Polyclonal | |
Unconjugated | |
THSD1 | |
4833423O18Rik; LOW QUALITY PROTEIN: thrombospondin type-1 domain-containing protein 1; thrombospondin type 1 domain containing 1; thrombospondin type I domain-containing 1 isoform 1; thrombospondin type-1 domain-containing protein 1; thrombospondin, type I, domain 1; thrombospondin, type I, domain containing 1; Thsd1; TMTSP; Transmembrane molecule with thrombospondin module; UNQ3010; UNQ3010/PRO9769 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55901 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NS62 | |
THSD1 | |
Synthetic peptide directed towards the middle region of human THSD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |