Learn More
Thrombopoietin/Tpo Mouse anti-Human, Clone: 34863, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2881
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13469755
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 27-03-2020
Description
Description
Thrombopoietin/Tpo Monoclonal antibody MAB2881 Specifically detects Thrombopoietin/Tpo in Human samples. It is validated for Western Blotting
Specifications
Specifications
Thrombopoietin/Tpo
34863
Western Blot 1 ug/mL
P40225
Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDF, MGDFC-mpl ligand, MKCSF, MK-CSF, ML, MPL ligand, MPLLG, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, THCYT1
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Thrombopoietin/Tpo in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse Tpo is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
THPO
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Thrombopoietin/Tpo Ser22-Gly353 Accession # P40225
Protein A or G purified from ascites
RUO
Primary
7066
Reconstitute at 0.5 mg/mL in sterile PBS.