missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombomodulin Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547550
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant human Thrombomodulin is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Thrombomodulin Polyclonal antibody specifically detects Thrombomodulin in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
Thrombomodulin | |
Polyclonal | |
Unconjugated | |
THBD | |
AHUS6; AI385582; BDCA3; BDCA-3; CD141; CD141 antigen; DC141; fetomodulin; sCD141; snoRNA MBII-339; soluble CD141; THBD; THPH12; THRM; thrombomdulin; thrombomodulin; thrombomodulin precursor; TM | |
Goat | |
Antigen affinity chromatography | |
RUO | |
21824 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P15306 | |
THBD | |
Mouse myeloma cell line NS0-derived recombinant mouse Thrombomodulin/BDCA-3 Leu17-Ser517 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |