Learn More
THOC6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543172
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: AGGDCQLHTM DLETGTFTRV LRGHTDYIHC LALRERSPEV LSGGEDGAVR Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 79%
THOC6 Polyclonal antibody specifically detects THOC6 in Human samples. It is validated for Western Blot.Specifications
THOC6 | |
Polyclonal | |
Unconjugated | |
THOC6 | |
BBIS; F830014G06Rik; fSAP35; Functional spliceosome-associated protein 35; hypothetical protein LOC450075; Pdrp; PSEC0006; THO complex 6; THO complex 6 homolog; THO complex 6 homolog (Drosophila); THO complex subunit 6 homolog; thoc6; WD repeat domain 58; WD repeat-containing protein 58; wdr58; zgc:101618 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79228 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86W42 | |
THOC6 | |
Synthetic peptide directed towards the middle region of human THOC6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |