missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Theophylline Monoclonal Antibody (TH11-84.4.1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110729
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-10729 detects Theophylline. MA1-10729 has been successfully used in ELISA applications. The MA1-10729 immunogen is Theophylline, conjugated to KLH.
Theophylline Monoclonal antibody specifically detects Theophylline in Chemical samples. It is validated for ELISA.Specifications
Theophylline | |
Monoclonal | |
∽1 mg/mL | |
PBS with no preservative; pH 7.4 | |
Mouse | |
purified | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA | |
TH11-84.4.1 | |
Unconjugated | |
C7H8N4O2; Dimethylxanthine; Theolair; Theophyllin | |
Theophylline, conjugated to KLH. | |
500 μg | |
Primary | |
Antibody | |
IgG1 |