Learn More
TGFBR3 Monoclonal Antibody (1C5H11), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517187
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA5-17187 targets TGFBR3 in indirect ELISA, WB applications and shows reactivity with Human and Mouse samples. The MA5-17187 immunogen is purified recombinant fragment of human TGFBR3 (amino acids: 147-328 ) expressed in E. Coli. MA5-17187 detects TGFBR3 which has a predicted molecular weight of approximately 93.4kDa.
TGFBR3 Monoclonal antibody specifically detects TGFBR3 in Human, Mouse samples. It is validated for ELISA, Western Blot.Specifications
TGFBR3 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
O88393, Q03167 | |
TGFBR3 | |
Purified recombinant fragment of human TGFBR3 (amino acids: 147-328 ) expressed in E. Coli. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
1C5H11 | |
Unconjugated | |
TGFBR3 | |
1110036H20Rik; AU015626; AW215636; betaglycan; betaglycan proteoglycan; BGCAN; OTTHUMP00000011787; TBRIII; tgf beta receptor 3; TGF-beta receptor type 3; TGF-beta receptor type III; Tgfbr3; TGFR-3; transforming growth factor beta receptor 3; transforming growth factor beta receptor III; Transforming growth factor beta receptor type 3; transforming growth factor, beta receptor 3; transforming growth factor, beta receptor III | |
Mouse | |
Protein G | |
RUO | |
21814, 7049 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |