Learn More
TGF-beta RII Rat anti-Mouse, Clone: 129502, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB532
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13450906
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 27-03-2020
Description
Description
TGF-beta RII Monoclonal antibody MAB532 Specifically detects TGF-beta RII in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
TGF-beta RII
129502
Western Blot 1 ug/mL
Q62312
AAT3, EC 2.7.11, EC 2.7.11.30, FAA3, HNPCC6, LDS1B, LDS2B, MFS2, RIIC, TAAD2, tbetaR-II, TGF-beta receptor type II, TGF-beta receptor type IIB, TGF-beta receptor type-2, TGF-beta type II receptor, TGFbetaRII, TGFbeta-RII, TGFBR2, TGF-bRII, TGFR-2, transforming growth factor beta receptor type IIC, transforming growth factor, beta receptor II (70/80kDa), transforming growth factor, beta receptor II (70/80kDa) isoform 1, transforming growth factor, beta receptor II (70/80kDa) isoform 2, transforming growth factor, beta receptor II (70-80kD), Transforming growth factor-beta receptor type II
Rat
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse TGF-beta RII in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with with recombinant rat MIS RII, recombinant mouse TGF-beta RI, recombinant human (rh) TGF-beta RII, rhTGF-beta RIIb, or rhTGF-beta RIII.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
TGFBR2
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse TGF-beta RII Ile24-Asp184 Accession # Q62312
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
7048
Reconstitute at 0.5 mg/mL in sterile PBS.