missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF alpha Sheep anti-Bovine, Human, Rat, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA185931
Code : LD
Additional Details : Weight : 0.01000kg
Description
Sheep anti Human TGF alpha antibody recognizes TGF-alpha (transforming growth factor alpha) from human, rat and bovine tissues and fluids.
TGF alpha (transforming growth factor alpha) is a 50 amino acid EGF related polypeptide expressed in a range of tissues, including skin, pancreas, liver, duodenum, salivary glands, urinary and respiratory systems, and the developing and mature central nervous system (CNS). TGF-alpha is produced by monocytes, keratinocytes and neuronal cells and stimulates the proliferation of a range of epidermal and epithelial cells, through finding to the epidermal growth factor receptor (EGFR). TGF alpha is mitogenic for multipotent neural progenitors and is involved in astrogliosis.Specifications
TGF alpha | |
Polyclonal | |
Unconjugated | |
P01135, P01134 | |
TGFA | |
Mature human recombit TGF-alpha conjugated to KLH | |
500 μg | |
Primary | |
Bovine, Human, Rat | |
Antibody | |
IgG |
ELISA | |
3.1 mg/mL | |
TGFA | |
TFGA | |
Sheep | |
Ion-exchange chromatography | |
RUO | |
24827, 540388, 7039 | |
4°C or -20°C if preferred | |
Liquid |