Learn More
TFPI Rat anti-Mouse, Clone: 372311, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2975
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13449825
  • £277.83 / 100µg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
TFPI Monoclonal antibody MAB2975 Specifically detects TFPI in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
TFPI
372311
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
TFPI
Mouse myeloma cell line NS0-derived recombinant mouse TFPI Leu29-Lys289 Accession # O54819
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
7035
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
O54819
convertin, EPI, EPITFPI1LACITFI, Extrinsic pathway inhibitor, LACI, Lipoprotein-associated coagulation inhibitor, tissue factor pathway inhibitor, tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor)
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse TFPI in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows no cross-reactivity with recombinant human (rh) TFPI or rmTFPI-2.