Learn More
TFPI-2 Mouse anti-Human, Clone: 243227, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB25451
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439525
  • £337.93 / 500µg
Estimated Shipment
28-06-2019
Description
Description
TFPI-2 Monoclonal antibody MAB25451 Specifically detects TFPI-2 in Human samples. It is validated for Immunoprecipitation, Neutralization
Specifications
Specifications
TFPI-2
243227
Immunoprecipitation 25 ug/mL, Neutralization 0.45 ug/mL
P48307
Placental protein 5, PP5, PP5FLJ21164, REF-1, TFPI2, TFPI-2REF1, tissue factor pathway inhibitor 2
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human TFPI-2 in direct ELISAs and Western blots.
Human
Immunoprecipitation, Neutralization
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
TFPI2
Mouse myeloma cell line NS0-derived recombinant human TFPI-2 Asp23-Phe235 Accession # P48307
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
7980
Reconstitute at 0.5 mg/mL in sterile PBS.