Learn More
TFF2 Mouse anti-Human, Clone: 366508, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB4077
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13440396
  • £288.60 / 100µg
Estimated Shipment
13-08-2020
Description
Description
TFF2 Monoclonal antibody MAB4077 Specifically detects TFF2 in Human samples. It is validated for Western Blotting
Specifications
Specifications
TFF2
366508
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
TFF2
E. coli-derived recombinant human TFF2 Glu24-Tyr129 Accession # Q03403
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
7032
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunohistochemistry
LYOPH
Western Blot 1 ug/mL, Immunohistochemistry 1-25 ug/mL
Q03403
SML1, SML1trefoil factor 2, SML1, human spasmolytic polypeptide (SP)10spasmolytic protein 1, SP, Spasmolysin, Spasmolytic polypeptide, trefoil factor 2
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human TFF2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human TFF3.