missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFB2M, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human TFB2M protein.
Brand: Abnova H00064216-B01P.50ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MWIPVVGLPRRLRLSALAGAGRFCILGSEAATRKHLPARNHCGLSDSSPQLWPEPDFRNPPRKASKASLDFKRYVTDRRLAETLAQIYLGKPSRPPHLLLECNPGPGILTQALLEAGAKVVALESDKTFIPHLESLGKNLDGKLRVIHCDFFKLDPRSGGVIKPPAMSSRGLFKNLGIEAVPWTADIPLKVVGMFPSRGEKRALWKLAYDLYSCTSIYKFGRIEVNMFIGEKEFQKLMADPGNPDLYHVLSVIWQLACEIKVLHMEPWSSFDIYTRKGPLENPKRRELLDQLQQKLYLIQMIPRQNLFTKNLTPMNYNIFFHLLKHCFGRRSATVIDHLRSLTPLDARDILMQIGKQEDEKVVNMHPQDFKTLFETIERSKDCAYKWLYDETLEDRSpecifications
TFB2M | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human TFB2M protein. | |
TFB2M | |
FLJ22661/FLJ23182/Hkp1 | |
Mouse | |
Affinity chromatography | |
RUO | |
64216 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_022366.1 | |
TFB2M | |
TFB2M (NP_071761.1, 1 a.a. ∼ 396 a.a) full-length human protein. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |