Learn More
TFAP2D Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540326
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: LGSSRPTPIL DLDIQRHLTH FSLITHGFGT PAICAALSTF QTVLSEMLNY Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
TFAP2D Polyclonal antibody specifically detects TFAP2D in Human samples. It is validated for Western Blot.Specifications
TFAP2D | |
Polyclonal | |
Unconjugated | |
TFAP2D | |
activating enhancer binding protein 2 beta-like 1; activating enhancer-binding protein 2 delta; activating enhancer-binding protein 2-delta; AP-2 like; AP-2-delta; AP2-delta; hypothetical protein LOC566838; si:dkey-5e16.2; Tcfap2d; TFAP2BL1; TFAP2D; transcription factor AP-2 beta (activating enhancer binding protein 2 beta)-like 1; transcription factor AP-2 beta (activating enhancer-binding protein 2 beta)-like 1; transcription factor AP-2 beta-like 1; transcription factor AP-2 delta; transcription factor AP-2 delta (activating enhancer binding protein 2 delta); transcription factor AP-2, delta; transcription factor AP-2-beta-like 1; Transcription factor AP-2-delta; zgc:114196 | |
Rabbit | |
Protein A | |
RUO | |
83741 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q7Z6R9 | |
TFAP2D | |
Synthetic peptide directed towards the C-terminal of human TFAP2D | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |