missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tex19.1 Monoclonal Antibody (678826), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA524308
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Tex19.1 Monoclonal antibody specifically detects Tex19.1 in Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
Tex19.1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q99MV2 | |
Tex19.1 | |
E. coli-derived recombinant mouse TEX19.1 Arg189-Pro351 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Immunocytochemistry, Western Blot | |
678826 | |
Unconjugated | |
Tex19.1 | |
2410081M02Rik; mTex19.1; testis expressed 19; testis expressed gene 19; testis expressed gene 19.1; Testis-expressed protein 19; Testis-expressed protein 19.1; testis-expressed protein 19A; testis-expressed sequence 19 protein; testis-expressed sequence 19A; Tex19; Tex19.1; Tex19a | |
Rat | |
Protein A/G | |
RUO | |
73679 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |