Learn More
UBIAD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA14378
Code : LD
Additional Details : Weight : 0.01000kg
Description
PA1-4378 detects the TERE1 protein in human samples. PA1-4378 has been successfully used in Western blot procedures. By Western blot this antibody detects a ∽37 kDa protein representing TERE1 in urothelial carcinoma. The PA1- 4378 immunogen is a synthetic peptide corresponding to residues P(32) E Q D R L P Q R S W R Q K(45) of human TERE1. There is 85% sequence identity to mouse and rat.
UBIAD1 Polyclonal antibody specifically detects UBIAD1 in Human samples. It is validated for Western Blot.Specifications
UBIAD1 | |
Polyclonal | |
Unconjugated | |
UBIAD1 | |
1200002M06Rik; AI426463; AW320947; RGD1309588; RP4-796F18.1; SCCD; Tere1; transitional epithelia response protein; transitional epithelial response protein 1; UbiA prenyltransferase domain containing 1; ubiA prenyltransferase domain-containing protein 1; UBIAD1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29914 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9Y5Z9 | |
UBIAD1 | |
Synthetic peptide corresponding to residues P(32) E Q D R L P Q R S W R Q K(45) of human TERE1. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |