missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEKT5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532004
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: PC-3, U87-MG, SK-N-SH. Predicted reactivity: Chimpanzee (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TEKT5 Polyclonal antibody specifically detects TEKT5 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
TEKT5 | |
Polyclonal | |
Unconjugated | |
TEKT5 | |
3300001K11Rik; CT149; RGD1307081; Tek5; Tekt5; tektin 5; Tektin5; tektin-5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
146279 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96M29 | |
TEKT5 | |
Recombinant fragment corresponding to a region within amino acids 1 and 227 of Human Tektin 5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |