Learn More
TCR V beta 10 Monoclonal Antibody (G101), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA180414
Code : LD
Additional Details : Weight : 0.01000kg
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Rat TCR V Beta 10 antibody, clone G101 recognizes the T-cell receptor V beta 10 gene product.
TCR V beta 10 Monoclonal antibody specifically detects TCR V beta 10 in Rat samples. It is validated for Flow Cytometry.Specifications
TCR V beta 10 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
0 | |
Tcrb | |
T cell clone expressing the TCR V beta 10 gene product. | |
250 μg | |
Primary | |
Rat | |
Antibody | |
IgG2a |
Flow Cytometry | |
G101 | |
Unconjugated | |
Tcrb | |
FLJ22602; MGC117436; MGC22624; MGC23964; MGC71411; RATTCB; RATTCBC1; T cell receptor beta locus; TCB; TCBC1; T-cell receptor beta chain; TCRA; TCRB; TCRD; TRA; TRB; TRB@ | |
Mouse | |
Ion-exchange chromatography | |
RUO | |
24820 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |