missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAC Monoclonal Antibody (3A8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen TCR1145
Code : PN
Additional Details : Weight : 0.80000kg
Description
TCR1145 targets TCR Alpha F1 in IHC (F), IHC (P) and IP applications and shows reactivity with Human samples. The TCR1145 immunogen is human TCR a chain constant region.
TRAC Monoclonal antibody specifically detects TRAC in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation.Specifications
TRAC | |
Monoclonal | |
0.15 mg/mL | |
PBS with 0.5% BSA and 0.1% sodium azide | |
P01848 | |
TRAC | |
Human TCR alpha chain constant region | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation | |
3A8 | |
Unconjugated | |
TRAC | |
IMD7; T-cell receptor; T-cell receptor alpha constant; TCRA; TRA; TRAC; TRCA | |
Mouse | |
Protein A | |
RUO | |
28755 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |