missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR alpha/beta Monoclonal Antibody (R73), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517541
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Store at 4°C; for long term storage, aliquot and freeze in single use volumes at -20°C, avoiding freeze-thaw cycles.
TCR alpha/beta Monoclonal antibody specifically detects TCR alpha/beta in Rat samples. It is validated for Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation.Specifications
TCR alpha/beta | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.02% sodium azide | |
0 | |
LOC290071, Tcrb | |
Rat T Blasts. | |
500 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation | |
R73 | |
Unconjugated | |
Tcrb | |
FLJ22602; IMD7; LOC290071; MGC117436; MGC22624; MGC23964; MGC71411; RATTCB; RATTCBC1; RGD1359684; similar to RIKEN cDNA A430107P09 gene; T cell receptor beta locus; T3/TCR complex; TCB; TCBC1; t-cell antigen receptor; T-cell receptor alpha constant; T-cell receptor beta chain; T-cell receptor V alpha; tcr alpha; TCR alpha/ beta; TCR beta; TCRA; Tcrb; TRA; Tra29; TRAC; TRB; TRB@; TRCA | |
Mouse | |
Protein G | |
RUO | |
24820, 290071 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |