missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCL1B Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547873
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, less than 5% cross-reactivity with recombinant human TCL1A is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
TCL1B Polyclonal antibody specifically detects TCL1B in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
TCL1B | |
Polyclonal | |
Unconjugated | |
TCL1B | |
Oncogene TCL1B; oncogene TCL-1B; SYN-1; Syncytiotrophoblast-specific protein; T-cell leukemia/lymphoma 1B; T-cell leukemia/lymphoma protein 1B; T-cell lymphoma/leukemia 1B; TCL1; TCL1/ MTCP1-like 1; TCL1/MTCP1-like protein 1; TCL1B; TML1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
9623 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O95988 | |
TCL1B | |
E. coli-derived recombinant human TCL1B Ala2-Asp128 (Glu93Arg) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |