missing translation for 'onlineSavingsMsg'
Learn More

TC10, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA11061

Code : LE

Additional Details : Weight : 0.01000kg

Product Code. 11585902

  • £292.00 / 100µg


Estimated Shipment
13-06-2018
Add to basket

Description

Description

PA1-1061 detects human and mouse TC10 protein in transfected cell samples and endogenous levels in tissue extracts. PA1-1061 is specific for isotype TC10a and demonstrates slight cross-reactivity with TC10b.PA1-1061 has successfully been used in immunoprecipitation and Western blot procedures. By Western blot, this antibody detects an ∽24kDa protein representing TC10 in transfected COS cells and a non-specific, unidentified band at ∽50 kDa.The PA1-1061 immunogen is a synthetic peptide corresponding D(136) P K T L A R L N D M K E K P I C(152) from human TC10. This peptide (Cat. # PEP-219) is available for use in neutralization and control experiments.
Specifications

Specifications

TC10
Polyclonal
Unconjugated
RHOQ
Purified
RHOQ
Synthetic peptide corresponding D(136) P K T L A R L N D M K E K P I C(152) from human TC10.
Antigen affinity chromatography
store at -20°C, Avoid Freeze/Thaw Cycles
Primary
23433
ImmunoprecipitationþWestern Blot
1mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
P17081, Q8R527
ARHQ, RASL7A, TC10, TC10A
Rabbit
IgG
100μg
Antibody
Polyclonal
HumanþMurine
Videos