missing translation for 'onlineSavingsMsg'
Learn More

TBX6, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA535065

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15387474

  • £304.30 / 400┬ÁL


Estimated Shipment
06-06-2018
Add to basket

Description

Description

This antibody is predicted to react with bovine and rat based on sequence homology.
Specifications

Specifications

TBX6
Polyclonal
PBS with 0.09% sodium azide
O95947, P70327
TBX6, T-box transcription factor TBX6
Rabbit
IgG
400μL
Antibody
Polyclonal
Murine
Western Blot
Unconjugated
TBX6
Purified
TBX6
KLH conjugated synthetic peptide between 143-172 amino acids from the central region of human TBX6
Size-exclusion - Dialysis, Ammonium sulfate precipitation
Store at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
6911
Videos