missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBX6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA535102
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine and rat based on sequence homology.
TBX6 Polyclonal antibody specifically detects TBX6 in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
TBX6 | |
Polyclonal | |
Unconjugated | |
TBX6 | |
rib-vertebrae; rv; SCDO5; T-box 6; T-box protein 6; T-box transcription factor 6; T-box transcription factor TBX6; TBX6 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
21389, 6911 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
O95947, P70327 | |
TBX6 | |
KLH conjugated synthetic peptide between 264-293 amino acids from the central region of human TBX6 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
Ig |