Learn More
TBR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534582
Code : NEW
Additional Details : Weight : 0.01000kg
Description
PA5-34582 targets TBR1 in ICC, IF, IHC (F), IHC (F), IHC (P), and WB applications and shows reactivity with Human, Mouse, and Rat samples. The PA5-34582 immunogen is synthetic peptide conjugated to KLH derived from within residues 50 - 150 of Mouse TBR1. PA5-34582 detects TBR1 which has a predicted molecular weight of approximately 74 kDa.
TBR1 Polyclonal antibody specifically detects TBR1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
TBR1 | |
Polyclonal | |
Unconjugated | |
TBR1 | |
LOW QUALITY PROTEIN: T-box brain protein 1; T-box brain gene 1; T-box brain protein 1; T-box brain transcription factor 1; T-box, brain 1; T-box, brain, 1; T-box, brain, 1-like; TBR1; TBR-1; T-brain-1; TES-56 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10716, 21375, 680427 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.0 mg/mL | |
PBS with 0.02% sodium azide; pH 7.4 | |
Q16650, Q64336 | |
TBR1 | |
Synthetic peptide conjugated to KLH derived from within residues 50 - 150 of Mouse TBR1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |